Epithelial Membrane Antigen (EMA) IHC Positive Control Slides, Newcomer Supply

Supplier: Newcomer Supply
3285A 3285B
75877-288BX 91.27 USD
75877-288 75877-290
Epithelial Membrane Antigen (EMA) IHC Positive Control Slides, Newcomer Supply
Slides Microscopy Control Slides
Epithelial Membrane Antigen (EMA) control slides contain sections of positive staining breast carcinoma and negative staining thyroid.

  • Tissue type: (+) breast / (-) Thyroid
  • All control tissue is sectioned on Superfrost® Plus slides
  • Staining validation technical memo included in each set
  • All tissues are formalin fixed, paraffin embedded FFPE
  • All slides are air dried
  • IHC slides are cut at 4 µm
  • Standard tissue placement is mid-high on slide

Newcomer supply Epithelial Membrane Antigen (EMA) control slides are for the positive immunohistochemical stainingMore Product Information
Order Now

SPECIFICATIONS

Species
Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR