Cytokeratin, HMW IHC Positive Control Slides, Newcomer Supply
Supplier: Newcomer Supply
3237A 3237B
75876-878BX 91.27 USD
75876-878 75876-880
Cytokeratin, HMW IHC Positive Control Slides, Newcomer Supply
Slides Microscopy Control Slides
Cytokeratin, HMW control slides contain sections of positive staining skin and negative staining myometrium.
  • Tissue type: (+) Skin / (-) Myometrium
  • All control tissue is sectioned on Superfrost® Plus slides
  • Staining validation technical memo included in each set
  • All tissues are formalin fixed, paraffin embedded FFPE
  • All slides are air dried
  • IHC slides are cut at 4 µm
  • Standard tissue placement is mid-high on slide
Newcomer supply cytokeratin, HMW (High Molecular Weight) control slides are for the positive immunohistochemical staining of Cytokeratin,More Product Information