CD30 IHC Positive Control Slides, Newcomer Supply
Supplier: Newcomer Supply
3175A 3175B
75876-858BX 91.27 USD
75876-858 75876-860
CD30 IHC Positive Control Slides, Newcomer Supply
Slides Microscopy Control Slides
CD30 control slides contain sections of positive staining Hodgkin’s lymphoma and negative staining kidney.
  • Tissue type: (+) Hodgkin's Lymphoma / (-) Kidney
  • All control tissue is sectioned on Superfrost® Plus slides
  • Staining validation technical memo included in each set
  • All tissues are formalin fixed, paraffin embedded FFPE
  • All slides are air dried
  • IHC slides are cut at 4 µm
  • Standard tissue placement is mid-high on slide
Newcomer supply CD30 control slides are for the positive immunohistochemical staining of CD30, expressed by Reed-Sternberg cells inMore Product Information