Formalex® GREEN Liquid Formalin Neutralizer for Sewer Disposal, Newcomer Supply
Supplier: Newcomer Supply
ABFX-01 ABFX-250 ABFX-05
75877-262EA 60.63 USD
75877-262 75877-264 75877-266
Formalex® GREEN Liquid Formalin Neutralizer for Sewer Disposal, Newcomer Supply
Spill Control Materials
Quickly and safely neutralize formalin waste for sewer disposal.
  • No sludgy residue
  • Contains no phosphates
  • No pH adjustment required
  • Non-vapor forming
  • Used in labs for over 27 years
Eco-friendly Formalex® “GREEN” Formalin Neutralizer effectively and easily neutralizes formalin waste for sewer disposal
1 gallon cube will neutralize 4 gallons of formalin waste
2.5 gallon cube will neutralize 10 gallons of formalin waste
5 gallon cube will neutralize 20 gallons of formalin waste
Simply add 32 ounces of Formalex® “GREEN” to each gallon of 10% neutralMore Product Information