Mast Cell, Animal Histo Positive Control Slides, Newcomer Supply

Supplier: Newcomer Supply
4412A 4412B
75877-316BX 75.55 USD
75877-316 75877-318
Mast Cell, Animal Histo Positive Control Slides, Newcomer Supply
Slides Microscopy Control Slides
Mast Cell, Animal control slides contain a section of positive staining animal tumor.

  • Tissue Type: (+) Animal Tumor
  • All control tissue is sectioned on Superfrost® Plus slides
  • Staining validation technical memo included in each set
  • All tissues are formalin fixed, paraffin embedded FFPE
  • All slides are air dried
  • Histo slides are cut at 4 µm, unless otherwise specified
  • Standard tissue placement is mid-high on slide

Newcomer supply Mast Cell, Animal control slides are for the positive histochemical staining of mast cells, cells filled with basophilic granules associated with inflammation and allergic reactions, which will stain metachromatically with toluidine blue.

Staining Results:
Mast cells will stain deep rose-violet.
Background will stain blue.

Ordering information: All controls are cut in serial sections.
10 set = 10 unstained slides and 1 stained slide.
98 set = 98 unstained slides and 2 stained slides. (First & Last Slides are Stained)
Reactivity of Histo control slides are guaranteed for one year from the date of receipt and should be revalidated after one year to verify continued reactivity.
Please note the following: In human tissue unless otherwise indicated. Histopathology control slides are not quality controlled for IHC methods, as such, we cannot guarantee antigenicity.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR