Luxol Fast Blue (LFB) Histo Positive Control Slides, Newcomer Supply
Supplier: Newcomer Supply
4407A 4407B
75877-048BX 75.55 USD
75877-048 75877-050
Luxol Fast Blue (LFB) Histo Positive Control Slides, Newcomer Supply
Slides Microscopy Control Slides
Luxol Fast Blue (LFB) control slides contain a section of positive staining spinal cord.
  • Tissue Type: (+) Spinal Cord
  • All control tissue is sectioned on Superfrost® Plus slides
  • Staining validation technical memo included in each set
  • All tissues are formalin fixed, paraffin embedded FFPE
  • All slides are air dried
  • Histo slides are cut at 8 µm, unless otherwise specified
  • Standard tissue placement is mid-high on slide
Newcomer supply Luxol Fast Blue (LFB) Control Slides are for the positive histochemical staining of myelin sheath in central nervous systemMore Product Information