Helicobacter sp. Histo Positive Control Slides, Newcomer Supply

Supplier: Newcomer Supply
4276A 4276B
75877-304BX 104.63 USD
75877-304 75877-306
Helicobacter sp. Histo Positive Control Slides, Newcomer Supply
Slides Microscopy Control Slides
Helicobacter sp. control slides contain sections of positive staining gastrointestinal tract and negative staining lung.

  • Tissue Type: (+) GI Tract
  • All control tissue is sectioned on Superfrost® Plus slides
  • Staining validation technical memo included in each set
  • All tissues are formalin fixed, paraffin embedded FFPE
  • All slides are air dried
  • Histo slides are cut at 4 µm, unless otherwise specified
  • Standard tissue placement is mid-high on slide

Newcomer supply Helicobacter sp. Control slides are for the positive histochemical staining of Helicobacter sp.,More Product Information
Order Now

SPECIFICATIONS

Species
Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR