Helicobacter sp., Artificial Histo Positive Control Slides, Newcomer Supply
Supplier: Newcomer Supply
4275A 4275B
75877-300BX 75.55 USD
75877-300 75877-302
Helicobacter sp., Artificial Histo Positive Control Slides, Newcomer Supply
Slides Microscopy Control Slides
Helicobacter sp., artificial control slides contain sections of positive staining rat lung and negative staining human lung.
  • Tissue Type: (+) Rat Lung
  • All control tissue is sectioned on Superfrost® Plus slides
  • Staining validation technical memo included in each set
  • All tissues are formalin fixed, paraffin embedded FFPE
  • All slides are air dried
  • Histo slides are cut at 4 µm, unless otherwise specified
  • Standard tissue placement is mid-high on slide
Newcomer supply Helicobacter sp., artificial control slides are for the positive histochemical staining of More Product Information