Cat Scratch, Artificial Histo Positive Control Slides, Newcomer Supply
Supplier: Newcomer Supply
4110A 4110B
75876-976BX 75.55 USD
75876-976 75876-978
Cat Scratch, Artificial Histo Positive Control Slides, Newcomer Supply
Slides Microscopy Control Slides
Cat scratch Bartonella sp., artificial control slides contain sections of positive staining rat lung and negative staining human lung.
  • Tissue Type: (+) Rat Lung / (-) Lung
  • All control tissue is sectioned on Superfrost® Plus slides
  • Staining validation technical memo included in each set
  • All tissues are formalin fixed, paraffin embedded FFPE
  • All slides are air dried
  • Histo slides are cut at 4 µm, unless otherwise specified
  • Standard tissue placement is mid-high on slide
Newcomer supply cat scratch Bartonella sp., artificial control slides are for theMore Product Information