Blastomyces sp., Animal Histo Positive Control Slides, Newcomer Supply
Supplier: Newcomer Supply
4091A 4091B
75876-968BX 75.55 USD
75876-968 75876-970
Blastomyces sp., Animal Histo Positive Control Slides, Newcomer Supply
Slides Microscopy Control Slides
Blastomyces sp., animal control slides contain sections of positive staining animal lymph node and negative staining human lung.
  • Tissue Type: (+) Animal Lymph Node / (-) Lung
  • All control tissue is sectioned on Superfrost® Plus slides
  • Staining validation technical memo included in each set
  • All tissues are formalin fixed, paraffin embedded FFPE
  • All slides are air dried
  • Histo slides are cut at 4 µm, unless otherwise specified
  • Standard tissue placement is mid-high on slide
Newcomer supply Blastomyces sp., animal control slides are for the positiveMore Product Information