Alcian Blue/PAS Histo Positive Control Slides, Newcomer Supply
Supplier: Newcomer Supply
4022A 4022B
75876-950BX 75.55 USD
75876-950 75877-292
Alcian Blue/PAS Histo Positive Control Slides, Newcomer Supply
Slides Microscopy Control Slides
Alcian blue/PAS control slides contain a section of positive staining small intestine.
  • Tissue Type: (+) small intestine
  • All control tissue is sectioned on Superfrost® Plus slides
  • Staining validation technical memo included in each set
  • All tissues are formalin fixed, paraffin embedded FFPE
  • All slides are air dried
  • Histo slides are cut at 4 µm, unless otherwise specified
  • Standard tissue placement is mid-high on slide
Newcomer supply alcian blue/PAS control slides are for the positive histochemical staining and differentiation of acidic epithelial mucinsMore Product Information