AdenoMag™ Transfection Reagent, OZ Biosciences
Supplier: OZ Biosciences
AdenoMag Transfection Reagent is a magnetic nanoparticles formulation specifically dedicated to enhance Adenovirus and Adeno Associated Virus (AAV) infection and transduction capacities.
- Increase transduction efficiency
- Accelerate the transduction process
- Efficient for hard-to-infect and non-permissive cells
- Concentrate the viral dose onto the cells
- Synchronise cells adsorption/infection without modifications of the viruses
Based on the Magnetofection™ technology, it allows concentrating the complete applied dose of viral particles onto cells within minutes, inducing a significant improvement of virus infectivity with extremely low vector doses.
Accessories information: This reagent needs to be used with a magnetic plate.
Delivery information: Shipped at room temperature
Packaging: Store at +4 °C