Lullaby™ Transfection Reagent, OZ Biosciences
Supplier: OZ Biosciences
LL70500 LL71000 LL73000
103258-114EA 231.03 USD
103258-114 103258-116 103258-118
Lullaby™ Transfection Reagent, OZ Biosciences
Transfection Reagents
Lullaby is the ideal siRNA transfection reagent for gene silencing
  • Effective with very low doses of siRNA
  • Minimized off-target effects
  • Suitable for siRNA, shRNA, dsRNA, etc.
  • Applicable to a broad range of cells
  • Serum compatible & non-toxic
  • Straightforward protocol
Relying on the TEE-technology, it has been successfully tested on numerous cell lines, reaching up to 90% gene silencing with high reproducibility and a very low toxicity.
Delivery information: Shipped at room temperatue
Packaging: Store at +4 °C