Eppendorf® 5920 R Refrigerated Large Centrifuges
Supplier: Eppendorf
5948000832 5948000131 5948000930 2231000842 5948000107 2231010155 5948000239
75874-302EA 20045.66 USD
75874-302 MP75874-302 75874-310 75874-306 75874-304 76443-014 75874-308
Eppendorf® 5920 R Refrigerated Large Centrifuges
Centrifuges Benchtop Centrifuges
The refrigerated Eppendorf Centrifuge 5920 R offers high-throughput performance and advanced temperature management in a very compact design. With a capacity of up to 4×1000 ml/52×50 ml conical tubes, it can spin both plates and tubes in the same bucket for real flexibility.
  • 'Centrifuge lid with soft-touch lid closure
  • Quiet operation to improve work environment
  • Compact footprint saves valuable bench space
  • Automatic rotor recognition and imbalance detection for maximum operational safety
  • Aerosol-tight Eppendorf QuickLock® caps and lids allow fast and ergonomic lid locking
  • Swing-bucket rotors and adapters accommodate tubes and bottles from 0.2 to 1000 ml
  • Plate rotor options for centrifugation of all types of MTP, PCR or Deepwell Plates
Fixed-angle rotors for high-speed molecular biology applications in tubes from 0.2 to 50 ml. FastTemp function for quick pre-cooling and FastTemp pro® function allows for automatic precooling based on pre-programmable time and date. Continuous cooling maintains a constant temperature after the run has been completed - samples stay cool.
ECO shut off engages after 8 hours (adjustable) of non-use to reduce energy consumption and to extend compressor life and Dynamic compressor control (DCC) technology for precise cooling performance.
The large bench top Centrifuge 5920 R combines extraordinary capacity with a compact and ergonomic product design. It has similar dimensions to competing refrigerated 3 L models yet it is capable of offering superior capacity of up to 4×1000 ml or 52×50 ml tubes. Streamline high-throughput centrifugation and make everyday work more enjoyable.
Universal buckets for both tubes and plates and save valuable time, money and space with universal buckets for both tubes and plates. With Rotor S-4×Universal-Large you can spin tubes, plates and bottles with only a change the adapter, not the rotor bucket. There is no need to exchange buckets, use separate bucket places or find space to store different buckets.
Superior 4 L capacity centrifuge 5920 R accommodates standard 1 L Nalgene bottles setting a new standard in the bench top centrifuge segment.The higher 4×1000 ml capacity allows you to process a higher volume without needing more runs.
Certifications: Certificate of Quality/Conformity (ISO 9001, ISO 13485, ISO 14001).