PUMPMATIC® Disposable Serological Pipettes, Polystyrene, Ashton Pumpmatic
Supplier: Ashton Pumpmatic
110 210 105 101 201
25470-002CS 723.56 USD
25470-002 25470-004 25470-008 25470-010 25470-000 25470-006 25470-012 25470-016
PUMPMATIC® Disposable Serological Pipettes, Polystyrene, Ashton Pumpmatic
Pipettes Serological Pipettes
PUMPMATIC® Disposable Serological Dispensing/Transfer Pipettes are available in sterile or nonsterile formats, with a variety of sizes and quantities to choose from.
  • Capacities available in 1, 5 and 10 ml
  • Accuracy ±0.02 %
  • Sterile and nonsterile
The Pumpmatic is a fast, cost-effective way to speed and simplify fluid sampling and transfers in any lab
Ideal for microbiology, blood banking, hematology, immunology, and urinalysis.
Each total closed Pumpmatic offers accurate, easy-to-read graduations, and the smooth action syringe make it easy toMore Product Information