Imidazole ≥99%, crystalline powder ACS
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Glyoxaline
, 1H-Imidazole
, Glyoxalamine
047274-36
047274-0E
047274-22
AA47274-22EA
55.62
USD
AA47274-22
AA47274-0E
AA47274-36
Imidazole ≥99%, crystalline powder ACS
Imidazole
Formula:
C₃H₄N₂ MW: 68.08 g/mol Boiling Pt: 256 °C Melting Pt: 88…92 °C Density: 1.03 Flash Pt: 145 °C (293 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00005183 CAS Number: 288-32-4 EINECS: 206-019-2 UN: 3263 ADR: 8,III Merck Index: 14,4912 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...