Water, liquid for LC-MS

Supplier: Thermo Scientific Chemicals

Synonyms: Dihydrogen oxide

047146-K2 047146-K7 047146-M6 047146-Y0
AA47146-K2EA 122.44 USD
AA47146-K2 AA47146-K7 AA47146-M6 AA47146-Y0
Water, liquid for LC-MS
Water
Formula: H₂O
MW: 18.02 g/mol
Boiling Pt: 100 °C
Melting Pt: 0 °C
Density: 1.000
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS Number: 7732-18-5
EINECS: 231-791-2
Merck Index: 14,10039

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR