Potassium hydroxide 45% (w/w) in aqueous solution, liquid

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Potash caustic, Potassium hydrate, Caustic potash

046072-AP 046072-K2 046072-K7
AA46072-APEA 44.17 USD
AA46072-AP AA46072-K2 AA46072-K7
Potassium hydroxide 45% (w/w) in aqueous solution, liquid
Potassium hydroxide

Colorless liquid

Formula: KOH
MW: 56.11 g/mol
Boiling Pt: 1320
Melting Pt: 360
Density: 1.456
Storage Temperature: Ambient
MDL Number: MFCD00003553
CAS Number: 1310-58-3
EINECS: 215-181-3
UN: 1814
ADR: 8 II
Merck Index: 12,07806

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR