Soybean oil, epoxidised, epoxidized
Supplier: SPECTRUM CHEMICAL MFG CORP
S1179-18KGBL
S1179-200LTB
S1179-500GM
S1179-2.5KG
700000-202EA
1643.22
USD
700000-202
700000-204
700000-206
700000-208
Soybean oil, epoxidised, epoxidized
Soybean oil, epoxidised
Soybean Oil, Epoxidized is used as a plasticizer and stabilizer in polyvinyl chloride (PVC) plastics.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Formula:
C₅₇H₉₈O₁₂ MW: 975.399 g/mol Density: 0.997 Storage Temperature: Ambient |
MDL Number:
MFCD00163560 CAS Number: 8013-07-8 |
Specification Test Results
Specific gravity at 25 °C | 0.988 - 0.998 |
Acid value | 0.50 |
Color (Alpha) | 125 |
Iodine value | 2.00 |
Moisture | 0.10% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...