PVC Peristaltic Pump Tubing, Glass Expansion
Supplier: Glass Expansion Inc
1.09WR 1.22RGRY 1.65BLU 2.79PW 0.13OBLK 0.57WY3B 0.95WBLK3B 1.02WHT 1.09WR3B 1.52YB 3.17BLKW 0.44GY 1.14RED 1.14RED3B 1.22RGRY3B 1.42YLW3B 2.54PO 0.19OR 0.19OR3B 0.38OG 0.44GY3B 0.51OY 0.51OY3B 0.64OW 0.95WBLK 1.85GRN 3.17BLKW3B 0.13OBLK3B 0.38OG3B 1.42YLW 1.52YB3B 1.85GRN3B 2.79PW3B 0.64OW3B 0.76BLK 0.89ORG 1.02WHT3B 0.25OB3B 0.57WY 0.89ORG3B 1.75BG 2.06PUR 2.06PUR3B 2.29PBLK3B 0.76BLK3B 1.65BLU3B 2.29PBLK 0.25OB 1.30GRY3B 2.54PO3B
101375-886PK 36.92 USD
101375-886 101375-906 101375-958 101376-030 101375-738 101375-808 101375-860 101375-866 101375-888 101375-938 101376-050 101375-786 101375-894 101375-896 101375-908 101375-928 101376-018 101375-746 101375-748 101375-766 101375-788 101375-794 101375-796 101375-814 101375-858 101375-974 101376-052 101375-740 101375-768 101375-926 101375-940 101375-976 101376-032 101375-816 101375-834 101375-846 101375-868 101375-756 101375-806 101375-848 101375-970 101375-986 101375-988 101376-008 101375-836 101375-960 101376-006 101375-754 101375-914 101376-020
PVC Peristaltic Pump Tubing, Glass Expansion
Tubing Transfer Tubing
Tubing for general laboratory applications.
  • Potential leaching of plasticizers
  • Does not age
  • Smooth inner surface with low gas permeability
  • Resistant to almost all inorganic chemicals
PVC two bridged tubing has 152mm between tags and three bridged has 7 mm between tags.
All tubing is made from Tygon R3603/R3607 or equivalent.