Sodium carbonate monohydrate Ph. Eur., NF, BP endotoxin tested
Supplier: Avantor Performance Materials
Warning
|
Synonyms:
Sodium carbonate hydrous
, Soda monohydrate
VWRVVWRB427-12KG
VWRB427-12KGEA
2017.06
USD
VWRB427-12KG
Sodium carbonate monohydrate Ph. Eur., NF, BP endotoxin tested
Sodium carbonate monohydrate
Tested according to the NF, EP, and BP monographs. Material is of non-animal, non-human origin.
- Bioprocessing
- Suitable for use as an Excipient
Formula:
Na₂CO₃·1H₂O MW: 124.00 g/mol Melting Pt: 107.85 °C Density: 2.25 g/cm³ (20 °C) |
MDL Number:
MFCD00149177 CAS Number: 5968-11-6 |
Specification Test Results
Assay (NF) | 99.5 – 100.5% |
Bacterial Endotoxins (NF) | ≤ 2.5 EU/g |
Heavy Metals |
≤ 10 ppm |
Identification A (NF) | PASS |
Identification B (NF) | PASS |
Water Determination | 12.0 – 15.0% |
Alkali Hydroxides and Bicarbonates | PASS |
Appearance of solution | PASS |
Arsenic (2.4.2 Method A) | ≤5 ppm |
Assay | 83.0 – 87.5% |
Chlorides (2.4.4) | ≤125 ppm |
Heavy Metals (2.4.8) | ≤50 ppm |
Identification A | PASS |
Identification B | PASS |
Identification C | PASS |
Iron (2.4.9) | ≤50 ppm |
Sulfates (2.4.13) | ≤250 ppm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...