EZ Wound Clips, Stoelting
Supplier: Stoelting
59020 59027
100228-840EA 647.31 USD
100228-840 100228-846
EZ Wound Clips, Stoelting
First Aid Supplies Wound Clips
This EZ Clip™ wound closing kit is perfect for rapidly closing wounds, providing maximum holding force with minimal trauma.
  • Easier and faster to use than conventional sutures and needles
  • After healing, the clips can be easily removed with the clip remover
  • Pyrogen-free and nonsterile
Delivery information: EZ Clip™ Kit includes the Applier, 9 mm wound clips (case of 1000) and the remover. Individual components may be ordered separately.
Packaging: Racks of 9 mm clips are individually packaged in heat sealed, peelaway envelopes for cleanliness andMore Product Information