Glassine Weighing Paper Sheets, Dyn-A-Med
Supplier: DYN-A-MED PRODUCTS
80047 80048 80049
490007-234PK 24.76 USD
490007-234 490007-236 490007-238
Glassine Weighing Paper Sheets, Dyn-A-Med
Weighing Papers
Glassine balance weighing paper for analytical weighing is designed for direct placement on any lab balance pan or for transferring material to another container.
  • Lightweight paper holds most analytical samples
  • Smooth, translucent surface will not trap material
  • Clean, dust-free sheets
  • Will not absorb moisture