dNTP Mix, 10 mM, BioChain®
Supplier: BIOCHAIN INSTITUTE INC MS
K6011105-1000 K6011105-100
100556-584EA 262.65 USD
100556-584 100558-414
dNTP Mix, 10 mM, BioChain®
Nucleic Acid Reagents Nucleotides
This dNTP reagent combines the highest quality with the most competitive price on the market.
  • Purified with preparative HPLC
  • Offers genomic DNA, RNA and cDNA
  • Convenient and economic source for large quantity PCR enzymes and reagents
This dNTP reagent combines the highest quality with the most competitive price on the market. The dNTPs are purified with preparative HPLC to achieve at least 99.5% purity. Also it offers genomic DNA, RNA and cDNA, a convenient and economic source for large quantity PCR enzymes and reagents.