Anti-MHC Class II (I-A/I-E) Rat Monoclonal Antibody (APC (Allophycocyanin)/Cy5.5®) [clone: M5/114.15.2]

Supplier: Tonbo Biosciences
65-5321-U025 65-5321-U100
75875-788EA 115.5 USD
75875-788 75875-790
Anti-MHC Class II (I-A/I-E) Rat Monoclonal Antibody (APC (Allophycocyanin)/Cy5.5®) [clone: M5/114.15.2]
Antibodies
The M5/114.15.2 antibody reacts with mouse MHC Class II alloantigens I-Ab, I-Ad, I-Aq, I-Ed, and I-Ek, as well as being cross-reactive with mouse cells of H-2p and H-2r haplotype. MHC Class II is widely expressed by mouse immune cells bearing these alloantigens, including T and B cells, monocytes, macrophages, and dendritic cells. The antibody does not react with the following alloantigens: I-Af, I-Ak, I-As, or NOD H-2g7. The M5/114.15.2 antibody may be used for analysis of mouse cells expressing MHC Class II alloantigens as described. Please note that the M5/114.15.2 clone may also be referred to as M5/114 in the literature.

Type: Primary
Antigen: MHC Class II (I-A/I-E)
Clonality: Monoclonal
Clone: M5/114.15.2
Conjugation: APC (Allophycocyanin)/Cy5.5®
Epitope:
Host: Rat
Isotype: IgG2b, kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR