Hydrochloric acid 0.5 M HCl (0.5N) in aqueous solution Reag. Ph. Eur. volumetric solution, Fluka™
Supplier: Honeywell Research Chemicals
|
Warning
|
Synonyms:
Hydrochloric acid solution, Muriatic acid
35329-10L-VP
35329-5L-VP
35329-1L
BJ35329-10L-VPEA
311.1
USD
BJ35329-10L-VP
BJ35329-5L-VP
BJ35329-1L
Hydrochloric acid 0.5 M HCl (0.5N) in aqueous solution Reag. Ph. Eur. volumetric solution, Fluka™
Hydrochloric acid
Formula:
HCl Boiling Pt: >100 °C (1.013 hPa) Melting Pt: 0 °C Density: 1.010 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011324 CAS Number: 7647-01-0 UN: 1789 ADR: 8,III |
Specification Test Results
Factor | 0.999 - 1.001 |
BAM/EMPA reference material | effective |
BAM/EMPA Certificate Number | effective |
Traceable to NIST SRM | effective |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...