Water Incubator_CO2

Supplier: GROWCELLS

Synonyms: Dihydrogen oxide

ISDW-1000 ISDW-2000 ISDW-010L ISDW-010Q ISDW-020Q
75831-945CS 474.92 USD
75831-945 75831-947 75831-949 75831-951 75831-953
Water Incubator_CO2
Water

Water for CO₂ Incubators is double distilled, 0.1um filter sterilized, has a pH of 7-9 with a conductivity of 1-20 microSiemens (Resistivity of 50K-1 M ohm) and is terminal sterilized. The water is free of all contaminants and exceeds the specifications set by CO₂ Incubator manufacturers.
Reusable & Re-autoclavable Polycarbonate Container.

Formula: H₂O
MW: 18.02 g/mol
Boiling Pt: 100
Melting Pt: 0
MDL Number: MFCD00011332
CAS Number: 7732-18-5
Merck Index: 13,10098

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR