VeriSpec™ Ytterbium Standard for ICP/MS 10 ppm in 2% HNO₃, RICCA Chemical Company
Supplier: RICCA Chemical Company
VeriSpec™
RV010636-50N RV010636-100N
RCRV010636100NEA 139.15 USD
RCRV010636100N RCRV010636-50N
VeriSpec™ Ytterbium Standard for ICP/MS 10 ppm in 2% HNO₃, RICCA Chemical Company
Standards Cationic Standards Ytterbium Standards
Certified reference material (CRM) for ICP-MS
Certifications: Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility