Anti-CD33 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: WM53]

Supplier: Southern Biotechnology
9590-11 9590-11S
101442-962EA 355.69 USD
101442-962 103265-382
Anti-CD33 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: WM53]
Antibodies
CD33 is a 67 kDa type I transmembrane glycoprotein and a member of the sialoadhesin family of cell surface receptors. It is absent from pluripotent stem cells but appears on myelomonocytic precursors after CD34. It then continues to be expressed on both the myeloid and monocyte lineages. While it has been reported that CD33 can function as a sialic acid-dependent cell adhesion molecule, cells expressing CD33 require desialylation before they can bind cells bearing the appropriate sialoglycoconjugates. This suggests that inhibitory cis interactions may regulate or block any adhesion function.

Type: Primary
Antigen: CD33
Clonality: Monoclonal
Clone: WM53
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR