Anti-IgG1 Goat Antibody (APC (Allophycocyanin))

Supplier: Southern Biotechnology
1070-11L 1070-11S
101442-630EA 643.38 USD
101442-630 101442-632
Anti-IgG1 Goat Antibody (APC (Allophycocyanin))
Antibodies
Goat Anti-Mouse IgG1-APC antibody with minimal reactivity to human proteins for use in flow cytometry assays.

Type: Secondary
Antigen: IgG1
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR