Anti-Mouse Kappa Goat Antibody

Supplier: Southern Biotechnology
1052-01
101442-626EA 238 USD
101442-626
Anti-Mouse Kappa Goat Antibody
Antibodies
Purified Goat F(ab’)2 Anti-Mouse Kappa antibody for use in ELISA, flow cytometry, immunohistochemistry, ELISpot, and stimulation assays.

Type: Secondary
Antigen: Kappa
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR