Anti-IgD Rat Antibody (RPE (R-Phycoerythrin)) [clone: 43430]

Supplier: Southern Biotechnology
1120-09L 1120-09
100242-776EA 377.92 USD
100242-776 100242-774
Anti-IgD Rat Antibody (RPE (R-Phycoerythrin)) [clone: 43430]
Antibodies
PE Anti-Mouse IgD antibody for use in flow cytometry assays.

Type: Secondary
Antigen: IgD
Clonality: Monoclonal
Clone: 43430
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR