Anti-IgG2c Goat Antibody (Biotin)

Supplier: Southern Biotechnology
1079-08
100242-670EA 295.54 USD
100242-670
Anti-IgG2c Goat Antibody (Biotin)
Antibodies
Goat Anti-Mouse IgG2c-Biotin antibody with minimal reactivity to human proteins for use in ELISA, flow cytometry, immunohistochemistry, and western blotting assays.

Type: Secondary
Antigen: IgG2c
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR