Anti-IgG (H+L) Goat Antibody (APC (Allophycocyanin))

Supplier: Southern Biotechnology
1031-11L 1031-11S
100242-514EA 311.23 USD
100242-514 100242-516
Anti-IgG (H+L) Goat Antibody (APC (Allophycocyanin))
Antibodies
Goat Anti-Mouse IgG(H+L)-APC antibody with minimal reactivity to human proteins for use in flow cytometry assays.

Type: Secondary
Antigen: IgG (H+L)
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR