Anti-CD5 Mouse Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: UCHT2]

Supplier: Southern Biotechnology
9526-09 9526-09S
100246-074EA 299.46 USD
100246-074 103260-198
Anti-CD5 Mouse Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: UCHT2]
Antibodies
CD5, a member of the scavenger receptor superfamily of cell surface receptors, is a type I transmembrane glycoprotein whose ligand is CD72. It is expressed on most thymocytes, all mature T cells, and a subset of B cells. CD5 plays a role in regulation of T cell activation and signaling.

Type: Primary
Antigen: CD5
Clonality: Monoclonal
Clone: UCHT2
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Rhesus, Cynomolgus, Chimpanzee
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR