Anti-CD9 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: MM2/57]

Supplier: Southern Biotechnology
9310-02 9310-02S
100245-872EA 299.46 USD
100245-872 103265-688
Anti-CD9 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: MM2/57]
Antibodies
CD9 is a 24 kDa member of the transmembrane 4 superfamily (TM4-SF) of cell surface receptors. It is expressed strongly on platelets, lymphoid progenitor cells, activated lymphocytes and endothelia, and, weakly, on eosinophils, granulocytes, monocytes, and macrophages. CD9 functions as an adhesion molecule where it mediates platelet aggregation and activation.

Type: Primary
Antigen: CD9
Clonality: Monoclonal
Clone: MM2/57
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Hamster, Canine, Feline, Monkey, Rabbit, Raccoon
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR