Anti-IgG (H+L) Goat Antibody (HRP (Horseradish Peroxidase))

Supplier: Southern Biotechnology
3050-05
100244-546EA 238 USD
100244-546
Anti-IgG (H+L) Goat Antibody (HRP (Horseradish Peroxidase))
Antibodies
Goat Anti-Rat IgG-HRP antibody with minimal reactivity to mouse proteins for use in ELISA, immunohistochemistry / immunocytochemistry, and western blotting assays.

Type: Secondary
Antigen: IgG (H+L)
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR