Anti-CD49b Rat Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: DX5]

Supplier: Southern Biotechnology
1806-09 1806-09L
100243-878EA 265.46 USD
100243-878 100243-880
Anti-CD49b Rat Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: DX5]
Antibodies
The monoclonal antibody DX5 reacts with CD49b, also known as very late antigen-2 (VLA-2) and α2 integrin. The antibody stains the majority of NK cells and a small subpopulation of T cells in all mouse strains tested (e.g., A/J, AKR, BALB/c, C3H/HeJ, C57BL/6, C57BL/10, C57BR, C58, CBA/Ca, CBA/J, DBA/1, DBA/2, SJL, SWR). Three-color flow cytometry analysis has revealed that most cells that express NK-1.1 (NKR-P1C) also express CD49b. However, small subsets of DX5+NK-1.1- and DX5-NK-1.1+ cells can be found, especially among the CD3+ population of T lymphocytes. NK cells cultured in the presence of IL-2 progressively lose reactivity with DX5 as a consequence of cellular proliferation. Therefore, DX5 can be used to define functionally distinct subsets of murine NK cells. The monoclonal antibody DX5 has not been demonstrated to have activating or blocking activity.

Type: Primary
Antigen: CD49b
Clonality: Monoclonal
Clone: DX5
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgM
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR