Anti-CD16/32 Rat Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: 93]

Supplier: Southern Biotechnology
1630-09L 1630-09
100243-436EA 371.38 USD
100243-436 100243-434
Anti-CD16/32 Rat Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: 93]
Antibodies
The lymphocyte Fcγ receptors recognize the Fc portion of IgG, presented either as immune complexes or as free antibody. The different classes of receptors are distinct because of varying size, tissue distribution and affinity for IgGs. The Fc type II receptor is expressed on a wide variety of cells including B cells, hematopoietic cells, monocyte/macrophages, neutrophils, platelets, Langerhans cells, eosinophils, basophils, trophoblasts, and endothelial cells of the placenta. The Fcγ type III receptors are higher affinity than the type II and are expressed on macrophages, NK cells and neutrophils. Both types of receptors can be expressed on the same cell and in varying ratios. The receptors are constitutively expressed, although cytokines and lymphokines can modulate their expression. Besides identifying FcγR+ cells, monoclonal antibodies to the FcγII/III receptor have been used to block Fc receptor binding of IgG, Fc-mediated signal transduction and effector functions, clearance of immune complexes and to attenuate infection by organisms dependent on FcγR for parasitic invasion. The monoclonal antibody 93 recognizes a conformational epitope formed by FcγII and FcγIII receptors.

Type: Primary
Antigen: CD16/32
Clonality: Monoclonal
Clone: 93
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR