Nutrient Agar with 1% Peptone, HiMedia
Supplier: HiMedia
M012-500G
M012-2.5KG
M012-5KG
95020-036EA
225.71
USD
95020-036
89222-190
89222-192
Nutrient Agar with 1% Peptone, HiMedia
Media for Microbiology
Dehydrated Media for Microbiology
Nutrient agar medium is used as a general purpose culture media.
- Cream to yellow homogeneous free flowing powder
- pH of 7.20 - 7.60
- Firm, comparable with 1.5 % Agar gel
Nutrient agar medium is used as a general purpose culture media which may be used as enriched medium by incorporating 10 % blood or other biological fluid.