Chemical Properties Kit
Supplier: AQUAPHOENIX SCIENTIFIC INC. SE
13-104
470200-082EA 278.85 USD
470200-082
Chemical Properties Kit
Educational Materials Chemistry Educational Materials Properties of Matter Learning Activities in Chemistry
Use Chemical Properties to Identify Unknown Solutions
  • Students identify unknowns from their own observations
  • Recommended for Grades 9-12
Students test the chemical properties of seven different solutions in every combination and identify chemical properties by observing the reactions. Evidence of chemical reactions observed include gas production, color changes, and formation of precipitates. After becoming familiar with the chemical properties of the solutions, students use the information to identify any of four unknown solutions.
Ordering information: Materials Include Sodium Bicarbonate, Hydrochloric Acid, Sodium Carbonate, Copper Sulfate, Ammonium Hydroxide, Phenolphthalein, Indicator Solution, Acetic Acid, Sodium Hydroxide, Unknowns A-D, Depression plates, Dropper Bottles, Labels.