Hydrochloric acid 37% ACS
Supplier: AQUA SOLUTIONS MS
|
|
Synonyms:
Muriatic acid
, Hydrochloric acid solution
H2505-500ML
H2505-2.5L
H2505-2.5L(PVC)
H2505-4L
101224-404EA
109.26
USD
101224-404
101223-072
101226-512
101223-970
Hydrochloric acid 37% ACS
Hydrochloric acid
Appearance Colorless fuming liquid
Harmonization Code 2806.10.0000
Formula:
HCl MW: 36.46 g/mol |
MDL Number:
MFCD00011324 CAS Number: 7647-01-0 UN: 1789 ADR: 8,II |
Specification Test Results
Assay | 36.5 – 38.0% |
Color (APHA) | Max. 10 |
Residue after Ignition | Max. 5 ppm |
Bromide (Br) | Max. 0.005% |
Free chlorine (Cl) | Max. 1 ppm |
Ammonium (NH4) | Max. 3 ppm |
Arsenic (As) | Max. 0.01 ppm |
Heavy metals (as Pb) | Max. 1 ppm |
Iron (Fe) | Max. 0.2 ppm |
Sulfate (SO4) | Max. 1 ppm |
Sulfite (SO2) | Max. 1 ppm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...