Screw Top Vials, 10 mm, PerkinElmer
Supplier: PerkinElmer
N9306057 N9306053 N9306057 N9306053
97013-862PK 69.64 USD
97013-862 CA97013-862 97013-858 CA97013-858
Screw Top Vials, 10 mm, PerkinElmer
Vials
Vials are manufactured from Type I borosilicate glass, which meets all USP, JP, and EP Pharmacopeia requirements.
  • Clear or amber color
  • Resistant to high temperature
  • Chemical resistant to acidic, neutral, and alkali solutions