SHOWA 377 Double Dipped Nitrile General-Purpose Work Glove, Showa
Supplier: Showa
377XL-09 377S-06 377L-08
89208-316CS 1210.05 USD
89208-316 89208-318 89208-310 89208-312 89208-314
SHOWA 377 Double Dipped Nitrile General-Purpose Work Glove, Showa
Gloves
An ideal glove for handling oily applications, with an additional layer of nitrile to prevent oil from reaching the hands.
  • Double dip nitrile to prevent oil from reaching the hands
  • Foam outer layer provides superior grip in oil
  • Extended cuff for added protection
Championed for its ultra-tactile grip and superior durability, this 13 gauge seamless knit glove features a full, thin, nitrile coating doubled with an absorbent foam nitrile palm coating and an extended cuff for added protection.
Ordering information: Order by Master case of 72 pair.
Packaging: Packaged 12 pair per poly bag, 6 bags per Master case.