ViviRen™ In-Vivo Renilla Luciferase Substrate, Promega
Supplier: Promega Corporation
P1231 P1232
PAP1231EA 203.23 USD
PAP1231 PAP1232
ViviRen™ In-Vivo Renilla Luciferase Substrate, Promega
Cell Culture Supplements and Additives
ViviRen in vivo Renilla Luciferase Substrate is a uniquely engineered coelenterazine-based compound with protected oxidation sites. These modifications are designed to minimize substrate degradation and autoluminescence.
  • Engineered Coelenterazine-Based Substrate for Animal Imaging
  • High-quality substrate eliminates potential interference in assays due to the presence of endotoxins
  • Available in multiple sizes to accommodate a variety of experimental settings
ViviRen in vivo Renilla Luciferase Substrate is a uniquely engineered coelenterazine-based compound with protected oxidation sites. These modifications are designed to minimize substrate degradation and autoluminescence. It is reported that the ViviRen Substrate demonstrates brighter output when compared to the native coelenterazine substrate when used in an in vivo imaging application in a mouse model. Cat.# P1231 is supplied as a liquid, 60mM in DMSO. Cat.# P1232 is supplied as a lyophilized solid.
In-Vivo Renilla Luciferase Substrate, 0.37 mg is supplied as a liquid60 mM in DMSO. In-Vivo Renilla Luciferase Substrate, 3.7 mg is supplied as a lyophilized solid.
The ViviRen Substrate demonstrates brighter output when compared to the native coelenterazine substrate when used in an in-vivo imaging application in a mouse model.