D-(-)-Luciferin potassium salt, VivoGlo™, in vivo grade

Supplier: Promega Corporation

P1041 P1042 P1043
PAP1041EA 259.66 USD
PAP1041 PAP1042 PAP1043
D-(-)-Luciferin potassium salt, VivoGlo™, in vivo grade
D(-)-Luciferin potassium salt

Firefly Luciferase Substrate for Use in Animal Imaging Studies.


  • High-quality substrate eliminates potential interference in assays due to the presence of endotoxins
  • Available in multiple sizes to accommodate a variety of experimental settings


Luciferase genes from the North American firefly (Photinus pyralis) and from other beetles are commonly used as light-emitting reporters in cellular and animal models. VivoGlo™ Luciferin is the potassium salt of d-luciferin, the firefly luciferase substrate capable of generating light when a suitable model is used. Product integrity is ensured by packaging most products in amber vials with septa as well as offering ease of dilution and use for imaging experiments. VivoGlo™ Luciferin is packaged with fine tolerances to minimize the need to weigh substrates.
Storage: -30°C to -10°C

Formula: C₁₁H₇KN₂O₃S₂
Storage Temperature: Freezer
MDL Number: MFCD00044928
CAS Number: 115144-35-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR