Gentamicin sulfate >590 µg/mg cell culture reagent

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: Gentamicin sulfate salt, NMD Inhibitor II, Garamycin, Gentamicin sulphate salt, Gentiomycin C

0219453001 0219453005
IC0219453001EA 160.25 USD
IC0219453001 IC0219453005
Gentamicin sulfate >590 µg/mg cell culture reagent
Gentamicin sulphate

Gentamicin is an aminoglycoside antibiotic complex produced by fermentation of Micromonospora purpurea or M. echinospora. It is a mixture of 3 major components designated as C1, C1a, and C2. Gentamicin is used as the sulfate salt. Each component consists of five basic nitrogens and requires five equivalents of sulfuric acid per mole of gentamicin base.


  • Storage temperature: +4 °C


Mode of action: Gentamicin causes codon misreading by binding to the 30S ribosomal subunit, blocking the translocation of peptidyl-tRNA from the acceptor site to the donor site.The bactericidal effect of gentamicin on Pseudomonas aeruginosa is exerted by the binding of gentamicin to the outer membrane, where it displaces natural cations, destabilizes the membrane, and forms holes in the cell surface. Antimicrobial spectrum: Gram-negative bacteria, Staphylococcus aureus and other Gram-positive bacteria.

Gentamicin sulfate is a broad spectrum antibiotic. It inhibits the growth of a wide variety of Gram-positive and Gram-negative microorganisms, including strains resistant to tetracycline, chloramphenicol, kanamycin, and colistin, particularly strains of Pseudomonas, Proteus, Staphylococcus, and Streptococcus. Gentamicin sulfate inhibits bacterial protein biosynthesis by binding to the 30S subunit of the ribosome.

Formula: C₂₁H₄₅N₅O₁₁S
Melting Pt: 218…237 °C (lit.)
MDL Number: MFCD00270181
CAS Number: 1405-41-0
EINECS: 215-778-9

Order Now

Specification Test Results

Physical Appearance White Powder
Optical Rotation +116 °
pH 3.5-5.5 (4% solution in water (Lit.))
Solubility Gentamicin sulfate is soluble in water (50 mg/mL), yielding a clear to very slightly hazy, colorless to faint. It is practically insoluble in alcohol and other organic solvents.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR