Heating Mantle, Buchner Funnel, Glas-Col®
Supplier: Glas-Col
100BTM810 100BTM816 100BTM820 100BTM814
33787-606EA 707.56 USD
33787-606 33787-610 33787-612 33758-200
Heating Mantle, Buchner Funnel, Glas-Col®
Heaters Heating Mantles
Heating for buchner funnel mantles.
  • Provides support for funnel
  • Warms solutions during transfer
  • Fabric interior hard shell exterior
Our buchner funnel heating mantle provides support for the funnel with gentle heating of solutions which need to be kept warm during transfer. Fabric interior softly nestles vessel to reduce the chance of thermal shock and damage. Withstands 400 degree C internal operating temperature.
Delivery information: 7-10 days after order.