Xylitol ≥99%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: (2R,3r,4S)-Pentane-1,2,3,4,5-pentaol

024311100GM
100268-822EA 137.31 USD
100268-822
Xylitol ≥99%
Xylitol
Formula: C₅H₁₂O₅
MW: 152.15 g/mol
Melting Pt: 92…96 °C
MDL Number: MFCD00064292
CAS Number: 87-99-0
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/58/75/21285875.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR