L-Valine-tert-butyl ester hydrochloride

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: H-Val-OtBu·HClL-2-Aminoisovaleric Acid tert-Butyl Ester Hydrochloride(S)-tert-Butyl 2-amino-3-methylbutanoate hydrochloride

0426765GM
100281-008EA 100.69 USD
100281-008
L-Valine-tert-butyl ester hydrochloride
L-Valine-tert-butyl ester hydrochloride
Formula: C₉H₁₉NO₂·HCl
MW: 209.7 g/mol
MDL Number: MFCD00034863
CAS Number: 13518-40-6
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/74/98/21287498.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR